Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzo[a]pyrene, Mouse, Clone: BAP-13, Abnova™

Catalog No. 89135469 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-469 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-469 Supplier Abnova Corporation Supplier No. MAB3600
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against Benzo[a]pyrene.

Specifications

Antigen Benzo[a]pyrene
Applications ELISA, Immunocytochemistry
Classification Monoclonal
Clone BAP-13
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against Benzo[a]pyrene.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Host Species Mouse
Immunogen Benzo[a]pyrene conjugated with BSA.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody is specific to Polynuclear Aromatic Hydrocarbons.This antibody BAP-13 inhibits DNA adduct formation (this ability was investigated in rat liver microsomes spiked with calf thymus DNA and 7,8-diol-B[a]P).
Target Species Bovine, Rat
Content And Storage Store at 4°C. Do not freeze.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.