Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BEST1, Mouse, Clone: E6-6, Abnova™

Catalog No. 89110936 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-936 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-936 Supplier Abnova Corporation Supplier No. MAB2330
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against synthetic peptide of BEST1.

This gene encodes a member of the bestrophin gene family. This small gene family is characterized by proteins with a highly conserved N-terminus with four to six transmembrane domains. Bestrophins may form chloride ion channels or may regulate voltage-gated L-type calcium-ion channels. Bestrophins are generally believed to form calcium-activated chloride-ion channels in epithelial cells but they have also been shown to be highly permeable to bicarbonate ion transport in retinal tissue. Mutations in this gene are responsible for juvenile-onset vitelliform macular dystrophy (VMD2), also known as Best macular dystrophy, in addition to adult-onset vitelliform macular dystrophy (AVMD) and other retinopathies. Alternative splicing results in multiple variants encoding distinct isoforms

Sequence: KDHMDPYWALENRDEAHS

Specifications

Antigen BEST1
Applications Immunofluorescence, Western Blot
Classification Monoclonal
Clone E6-6
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic peptide of BEST1.
Dilution Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In buffer containing 0.09% sodium azide
Gene BEST1
Gene Alias ARB/BEST/BMD/TU15B/VMD2
Gene Symbols BEST1
Host Species Mouse
Immunogen A synthetic peptide (conjugated with KLH) corresponding to human BEST1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7439
Test Specificity This antibody recognizes a band at ∼68 KDa on Western Blot.
Target Species Human, Pig, Primate
Content And Storage Store at -20°C or -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.