Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Mouse anti-Betacellulin, Clone: 6G10.1,

Catalog No. MABS458MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABS458MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABS458MI Supplier MilliporeSigma™ Supplier No. MABS458
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects Betacellulin clone: 6G10.1 in Human samples, and it is validated for Immunohistochemistry, Western Blotting

The human BTC gene (Gene ID 685) encodes the 178 aa preprobetacellulin (UniProt P35070) protein. Post-translatioinal removal of the signal peptide (aa 1-31) produces probetacellulin (aa 32-178), which further matures into the 80 aa betacellulin (aa 32-111) following propeptide (aa 112-178) removal. Betacellulin is an EGF family growth factor with a higher affinity and specificity for ErbB1/EGFR and ErbB4. Betacellulin plays an important role in the development of the acute exacerbations of chronic obstructive pulmonary disease. Betacellulin is also reported to induce CXCL8 (IL-8) production from human lung cancer cells through EGFR–PI3K–Akt–ERK pathway activation, resulting in the recruitment of inflammatory cells in the cancer tissue and creating an inflammatory tumor microenvironment.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Betacellulin
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 6G10.1
Formulation Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. P35070
Gene Symbols BTC
Host Species Mouse
Immunogen GST-tagged human recombinant full-length mature betacellulin.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_001720
Target Species Human
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.