Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BFSP1, Mouse, Clone: FIL-27, Abnova™

Catalog No. 89111002 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-002 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-002 Supplier Abnova Corporation Supplier No. MAB2423
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native BFSP1.

Specifications

Antigen BFSP1
Applications Immunohistochemistry (Frozen), Western Blot
Classification Monoclonal
Clone FIL-7B10
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native BFSP1.
Dilution Western Blot (1-2 ug/mL) Immunohistochemistry (Frozen sections) (2-4 ug/mL) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from 1.2% sodium acetate (2mg BSA, 0.01mg sodium azide)
Gene Accession No. BC041483
Gene Alias CP115/CP94/FILENSIN/LIFL-H
Gene Symbols BFSP1
Host Species Mouse
Immunogen Native from human and bovine lens filament enriched fraction (plasma membrane-cytoskeleton complex).
Purification Method Affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 631/281644
Target Species Bovine, Human
Content And Storage Store at -20°C on dry atmosphere. After reconstitution with 1mL of 1.2% sodium acetate or neutral PBS to a concentration of 100μg/mL store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.