Learn More
BFSP1, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant BFSP1.
Supplier: Abnova Corporation H00000631A01
Description
More than 99% of the vertebrate ocular lens is comprised of terminally differentiated lens fiber cells. Two lens-specific intermediate filament-like proteins, CP49 (also known as phakinin) and the protein product of this gene, filensin, are expressed only after fiber cell differentiation has begun. Both proteins are found in a structurally unique cytoskeletal element that is referred to as the beaded filament (BF). Mutations in this gene are the cause of autosomal recessive cortical juvenile-onset cataract. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Sequence: EESRRPCAMVTPGAEEPSIPEPPKPAADQDGAEVLGTRSRSLPEKGPPKALAYKTVEVVESIEKISTESIQTYEETAVIVETMIGKTKSDKKKSGEKSSpecifications
BFSP1 | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant BFSP1. | |
BFSP1 | |
CP115/CP94/FILENSIN/LIFL-H | |
Mouse | |
50 μL | |
Primary | |
Human | |
Antisera |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_001195 | |
BFSP1 | |
BFSP1 (NP_001186, 567 a.a. to 664 a.a) partial recombinant protein with GST tag. | |
RUO | |
631 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only