Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BGLAP, Mouse, Clone: 2H9, Abnova™

Catalog No. 89111192 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-192 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-192 Supplier Abnova Corporation Supplier No. MAB2987

Mouse monoclonal antibody raised against BGLAP.

Specifications

Antigen BGLAP
Applications ELISA
Classification Monoclonal
Clone 2H9
Conjugate Unconjugated
Description bone gamma-carboxyglutamate (gla) protein
Formulation In 0.9% NaCl (0.09% sodium azide)
Gene BGLAP
Gene Alias BGP, OC, PMF1
Gene Symbols BGLAP
Host Species Mouse
Immunogen A mixture of bovine BGLAP (conjugated with KLH) and recombinant GST fusion protein corresponding to human BGLAP.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 632
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.