Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BGLAP, Mouse, Clone: OCG3, Abnova™

Catalog No. 89113451 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-451 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 89-113-451 Supplier Abnova Corporation Supplier No. MAB8377
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse monoclonal antibody raised against BGLAP.

Specifications

Antigen BGLAP
Applications Immunohistochemistry, Immunohistochemistry (Frozen), ELISA, Western Blot
Classification Monoclonal
Clone OCG3
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against BGLAP.
Dilution Sandwich ELISA (1-10 ug/mL) Western Blot (1-10 ug/mL) Immunohistochemistry (Frozen sections) (1-10 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1-10 ug/mL) The optimal working dilution should be determined by th
Formulation Lyophilized from 10mM PBS, pH 7.4 (1% BSA, 0.09% sodium azide)
Gene BGLAP
Gene Symbols BGLAP
Host Species Mouse
Immunogen Bovine BGLAP.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 281646
Target Species Bovine
Content And Storage Store at 4°C on dry atmosphere. After reconstitution with deionized waterstore at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.