Learn More
BID, Mouse, Clone: 3C5, Abnova™
Mouse monoclonal antibody raised against partial recombinant BID.
Supplier: Abnova Corporation MAB10503
Description
This gene encodes a death agonist that heterodimerizes with either agonist BAX or antagonist BCL2. The encoded protein is a member of the BCL-2 family of cell death regulators. It is a mediator of mitochondrial damage induced by caspase-8 (CASP8); CASP8 cleaves this encoded protein, and the COOH-terminal part translocates to mitochondria where it triggers cytochrome c release. Multiple alternatively spliced transcript variants have been found, but the full-length nature of some variants has not been defined. [provided by RefSeq
Specifications
BID | |
Monoclonal | |
Unconjugated | |
ELISA (1:10000) Western Blot (1:500-1:2000) Immunohistochemistry (1:200-1:1000) Immunofluorescence (1:200-1:1000) Flow cytometry (1:200-1:400) The optimal working dilution should be determined by the end user. | |
BID | |
BID | |
Recombinant protein corresponding to human BID. | |
RUO | |
637 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG1 |
ELISA, Flow Cytometry, Immunofluorescence, Immunohistochemistry, Western Blot | |
3C5 | |
Mouse monoclonal antibody raised against partial recombinant BID. | |
In ascites (0.03% sodium azide) | |
FP497/MGC15319/MGC42355 | |
Mouse | |
100 μL | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.