Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BLK, Mouse, Clone: 9D10B7, Abnova™

Catalog No. 89108639 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-108-639 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-108-639 Supplier Abnova Corporation Supplier No. MAB10453
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant BLK.

Specifications

Antigen BLK
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 9D10B7
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant BLK.
Dilution Western Blot (1:500-1:2000) Immunohistochemistry (1:200-1:1000) ELISA (1:10000) The optimal working dilution should be determined by the end user.
Formulation In ascitic (0.03% sodium azide)
Gene BLK
Gene Alias MGC10442
Gene Symbols BLK
Host Species Mouse
Immunogen Recombinant protein corresponding to human BLK.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 640
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.