Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Blood Group A Mouse anti-Human, Alexa Fluor 647, Clone: NaM87-1F6, BD

Catalog No. BDB565384 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB565384 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB565384 Supplier BD Biosciences Supplier No. 565384
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The NaM87-1F6 monoclonal antibody specifically binds to blood group A expressed on human erythrocytes. Clone NaM87-1F6 induces agglutination of human group A red blood cells. It does not react with human group B, or group O red blood cells.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Blood Group A
Applications Flow Cytometry
Classification Monoclonal
Clone NaM87-1F6
Conjugate Alexa Fluor 647
Formulation Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG3 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.