Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Blood Group A, Mouse, Clone: 9A, Abnova™

Catalog No. 89136333 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-333 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-333 Supplier Abnova Corporation Supplier No. MAB7313
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against Blood Group A Antigen.

Specifications

Antigen Blood Group A
Applications ELISA, Immunoprecipitation
Classification Monoclonal
Clone 9A
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against Blood Group A Antigen.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS
Host Species Mouse
Immunogen Native purified from human vulval squamous carcinoma cell line (A431 cells).
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity 9A is suitable for binding to the EGF receptor in A431 cells only. These cells have a glycosylation defect which reveals otherwise cryptic sugar structures.
Target Species Human
Content And Storage Store at 4°C for short term. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.