Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Blood group A1B, Mouse, Clone: HE-24, Abnova™
Mouse monoclonal antibody raised against partial native Blood group antigen A1B.
Supplier: Abnova Corporation MAB5325
Specifications
| Blood group A1B | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| Mouse | |
| 1 mL | |
| Primary | |
| Human | |
| Liquid |
| Agglutination, Immunohistochemistry | |
| HE-24 | |
| Mouse monoclonal antibody raised against partial native Blood group antigen A1B. | |
| In culture supernatant | |
| Native purified from glycoprotein fraction isolated from saliva of secretors with blood group A. | |
| RUO | |
| This antibody distinguishes blood group A1B from A2B. The specifity of This antibody HE-24 was verified on >1000 samples of blood. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgM |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction