Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Blood Group ABH, Mouse, Clone: HE-10, Abnova™

Catalog No. 89136321 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-321 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-321 Supplier Abnova Corporation Supplier No. MAB7176

Mouse monoclonal antibody raised against Blood Group ABH.

Specifications

Antigen Blood Group ABH
Applications Immunohistochemistry
Classification Monoclonal
Clone HE-10
Conjugate Unconjugated
Formulation In tissue culture supernatant
Host Species Mouse
Immunogen A mixture of erythrocytes of blood group A1 and glycoprotein fraction isolated from saliva of secretors with blood group A.
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.