Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Blood Group Lewis a, Mouse, Clone: 7LE, Abnova™

Catalog No. 89135468 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-468 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-468 Supplier Abnova Corporation Supplier No. MAB3592
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native Blood Group Lewis a .

Specifications

Antigen Blood Group Lewis a
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 7LE
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native Blood Group Lewis a .
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Host Species Mouse
Immunogen Native purified human Blood Group Lewis a.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody recognizes Lewisa blood group antigen, a carbohydrate determinantcarried on both glycolipids and glycoproteins, expressed in colonic epithelial cells. Lewisamay be useful for detection of gastrointestinal, pancreatic and colorectal tumors.
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.