Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Mouse anti-Bmi-1, Clone: F6, Alexa Fluor 488,

Catalog No. 05637AF488 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Alexa Fluor 488
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
05-637-AF488 100 μL Alexa Fluor 488
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-637-AF488 Supplier MilliporeSigma™ Supplier No. 05637AF488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects Bmi-1 clone: F6 in Human, Monkey samples, and it is validated for Immunocytochemistry

Polycomb complex protein BMI-1 (UniProt P25916; also known as B lymphoma Mo-MLV insertion region 1, Polycomb group RING finger protein 4) is encoded by the Bmi1 (also known as AW546694, Bmi-1, Pcgf4) gene (Gene ID 12151) in murine species. Bmi1 is a member of the polycomb group (PcG) family of evolutionarily conserved gene silencers involved in cell fate decisions and body pattern during development. Bmi1 is an important regulator of self-renewal of hematopoietic and neural stem cells, as well as cancer stem cells (CSCs). Bmi1 is a main constituent of the polycomb repressive complex 1 (PRC1) and pariticipates in PRC-mediated gene silencing by promoting monoubiquitination of H2A. Bmi1 overexpression is reported in many different human cancers and high level of Bmi1 is considered as a poor prognostic marker. Bmi1 targets genes involved in transforming growth factor-β/bone morphogenetic protein (TGF-beta/BMP), endoplasmic reticulum (ER) stress, and WNT pathways. Bmi itself is transcriptionally regulated by c-Myc and E2F1 transcription factors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Bmi-1
Applications Immunocytochemistry
Classification Monoclonal
Clone F6
Conjugate Alexa Fluor 488
Formulation Purified mouse conjugate antibody in PBS with 15mg/mL BSA and 0.1% sodium azide.
Gene Accession No. P25916
Gene Symbols BMI1; PCGF4; RNF51
Host Species Mouse
Immunogen Recombinant Bmi-1 protein corresponding to residues 1-202 of mouse Bmi-1.
Purification Method Protein G Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_031578.2
Target Species Human, Monkey
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.