Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BMP2K, Mouse, Clone: 4B19, Abnova™

Catalog No. 89107046 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-046 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-046 Supplier Abnova Corporation Supplier No. H00055589M04
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant BMP2K.

This gene is the human homolog of mouse BMP-2-inducible kinase. Bone morphogenic proteins (BMPs) play a key role in skeletal development and patterning. Expression of the mouse gene is increased during BMP-2 induced differentiation and the gene product is a putative serine/threonine protein kinase containing a nuclear localization signal. Therefore, the protein encoded by this human homolog is thought to be a protein kinase with a putative regulatory role in attenuating the program of osteoblast differentiation. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Sequence: YQQAFFQQQMLAQHQPSQQQASPEYLTSPQEFSPALVSYTSSLPAQVGTIMDSSYSANRSVADKEAIANFTNQKNISNPPDMSGWNPFGEDNFSKLTEEELLDREFDLLRS

Specifications

Antigen BMP2K
Applications ELISA, Western Blot
Classification Monoclonal
Clone 4B19
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant BMP2K.
Formulation PBS with no preservative; pH 7.4
Gene BMP2K
Gene Accession No. BC036021
Gene Alias BIKE/DKFZp434K0614/DKFZp434P0116/HRIHFB2017
Gene Symbols BMP2K
Host Species Mouse
Immunogen BMP2K (AAH36021,540 a.a. ∽ 650 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Protein A Purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55589
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.