Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bmp4, Mouse, Clone: 3H2, Abnova™

Catalog No. 89111444 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-444 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-444 Supplier Abnova Corporation Supplier No. MAB3491

Mouse monoclonal antibody raised against recombinant Bmp4.

Specifications

Antigen Bmp4
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 3H2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant Bmp4.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:10-1:20) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from tissue culture supernatant (0.09% sodium azide)
Gene Bmp4
Gene Alias Bmp2b/Bmp2b-1/Bmp2b1
Gene Symbols Bmp4
Host Species Mouse
Immunogen Recombinant protein corresponding to mouse Bmp4.
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12159
Target Species Frog, Human, Mouse
Content And Storage Store at 4°C on dry atmosphere. After reconstitution with distilled water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.