Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BMPR1A, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89104162 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-104-162 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-104-162 Supplier Abnova Corporation Supplier No. H00000657A03
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length recombinant BMPR1A.

The bone morphogenetic protein (BMP) receptors are a family of transmembrane serine/threonine kinases that include the type I receptors BMPR1A and BMPR1B and the type II receptor BMPR2. These receptors are also closely related to the activin receptors, ACVR1 and ACVR2. The ligands of these receptors are members of the TGF-beta superfamily. TGF-betas and activins transduce their signals through the formation of heteromeric complexes with 2 different types of serine (threonine) kinase receptors: type I receptors of about 50-55kDa and type II receptors of about 70-80kDa. Type II receptors bind ligands in the absence of type I receptors, but they require their respective type I receptors for signaling, whereas type I receptors require their respective type II receptors for ligand binding. [provided by RefSeq

Sequence: MPQLYIYIRLLGAYLFIISRVQGQNLDSMLHGTGMKSDSDQKKSENGVTLAPEDTLPFLKCYCSGHCPDDAINNTCITNGHCFAIIEEDDQGETTLASGCMKYEGSGFQCKDSPKAQLRRTIECCRTNLCNQYLQPTLPPVVIGPFFDGSIRWLVLLISMAVCIIAMIIFSSCFCYKHYCKSISSRRRYNRDLEQDEAFIPVGESLKDLVDQSQSSGSGSGLPLLVQRTIAKQIQMVRQVGKGRYGEVWMGKWRGEKVAVKVFFTTEEASWFRETEIYQTVLMRHENILGFIAADIKGTGSWTQLYLITDYHENGSLYDFLKCATLDTRALLKLAYSAACGLCHLHTEIYGTQGKPAIAHRDLKSKNILIKKNGSCCIADLGLAVKFNSDTNEVDVPLNTRVGTKRYMAPEVLDESLNKNHFQPYIMADIYSFGLIIWEMARRCITGGIVEEYQLPYYNMVPSDPSYEDMREVVCVKRLRPIVSNRWNSDECLRAVLKLMSECWAHNPASRLTALRIKKTLAKMVESQDVKI

Specifications

Antigen BMPR1A
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length recombinant BMPR1A.
Formulation 50% glycerol
Gene BMPR1A
Gene Accession No. BC028383
Gene Alias 10q23del/ACVRLK3/ALK3/CD292
Gene Symbols BMPR1A
Host Species Mouse
Immunogen BMPR1A (AAH28383, 1 a.a. to 532 a.a) full-length recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 657
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.