Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bovine rotavirus, Mouse, Clone: 3C10, Abnova™

Catalog No. 89134922 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-134-922 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-134-922 Supplier Abnova Corporation Supplier No. MAB1289
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against bovine rotavirus.

Specifications

Antigen Bovine rotavirus
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 3C10
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against bovine rotavirus.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.1% gelatin, 0.09% sodium azide)
Host Species Mouse
Immunogen Native purified bovine rotavirus.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody reacts with rotavirus p42 antigen. It reacts with monkey rotavirus (SA-11), porcine rotavirus (PP) and human rotaviruses (field strains).
Content And Storage Store at -20°C. This antibody can be stored at 4°C for short term storage.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.