Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Mouse anti-BPTF, Clone: 15B1.1,

Catalog No. MABE443MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE443MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE443MI Supplier MilliporeSigma™ Supplier No. MABE443
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects BPTF clone: 15B1.1 in Human, Mouse, Rat samples, and it is validated for Immunohistochemistry (Paraffin), Western Blotting

Nucleosome-remodeling factor (NURF) subunit BPTF (UniProt E9Q6A7 & A2A654; also known as Bromodomain and PHD finger-containing transcription factor, Fetal Alzheimer antigen) is encoded by the Bptf (also known as 9430093H17Rik, Falz) gene (Gene ID 207165) in murine species. ATP-dependent chromatin remodeling complexes are classified into four major subfamilies (SWI/SNF, ISWI, CHD, and INO80). NURF is the founding member of ISWI remodeling complexes. Mammalian NURF is composed of BPTF, the ATPase SNF2L, and the Trp-Asp (WD) repeat protein pRBAP46/48, with BPTF being the largest subunit among the three. BPTF PHD finger-mediated binding of H3K4me3 stabilizes the NURF complex on chromatin, resulting in NURF-catalyzed nucleosome sliding. The NURF remodeling complex is essential for mammalian development and executes epigenetic regulation in multiple signaling pathways, including heat shock, TGF-beta/SMAD, JAK/STAT, WNT/beta-catenin, RB, and nuclear hormone receptors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BPTF
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 15B1.1
Formulation Purified mouse monoclonal IgG2bκ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. E9Q6A7
Gene Symbols Bptf
Host Species Mouse
Immunogen GST-tagged recombinant protein corresponding to mouse BPTF.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_789820
Target Species Human, Mouse, Rat
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.