Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRCA1 Mouse anti-Human, Monoclonal Antibody, Abnova™

Catalog No. 89110866 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-866 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-866 Supplier Abnova Corporation Supplier No. MAB2170
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant BRCA1.

This gene encodes a nuclear phosphoprotein that plays a role in maintaining genomic stability, and it also acts as a tumor suppressor. The encoded protein combines with other tumor suppressors, DNA damage sensors, and signal transducers to form a large multi-subunit protein complex known as the BRCA1-associated genome surveillance complex (BASC). This gene product associates with RNA polymerase II, and through the C-terminal domain, also interacts with histone deacetylase complexes. This protein thus plays a role in transcription, DNA repair of double-stranded breaks, and recombination. Mutations in this gene are responsible for approximately 40% of inherited breast cancers and more than 80% of inherited breast and ovarian cancers. Alternative splicing plays a role in modulating the subcellular localization and physiological function of this gene. Many alternatively spliced transcript variants, some of which are disease-associated mutations, have been described for this gene, but the full-length natures of only some of these variants has been described. A related pseudogene, which is also located on chromosome 17, has been identified. [provided by RefSeq

Specifications

Antigen BRCA1
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant BRCA1.
Dilution Western Blot (1-10 ug/mL) Immunoprecipitation (1-10 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4
Gene BRCA1
Gene Alias BRCAI/BRCC1/IRIS/PSCP/RNF53
Gene Symbols BRCA1
Host Species Mouse
Immunogen Recombinant GST fusion protein corresponding to amino acids 341-748 of BRCA1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 672
Test Specificity This antibody recognizes full-length BRCA1, a 220 kD nuclear phosphoprotein.
Target Species Human
Content And Storage Store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.