Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRCC3, Mouse, Clone: AT3B1, Abnova™

Catalog No. 89113181 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-181 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-181 Supplier Abnova Corporation Supplier No. MAB7555
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant BRCC3.

This gene encodes a subunit of the BRCA1-BRCA2-containing complex (BRCC), which is an E3 ubiquitin ligase. This protein is also thought to be involved in the cellular response to ionizing radiation and progression through the G2/M checkpoint. Alternative splicing results in multiple transcript variants. [provided by RefSeq

Specifications

Antigen BRCC3
Applications ELISA, Western Blot
Classification Monoclonal
Clone AT3B1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant BRCC3.
Dilution Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene BRCC3
Gene Alias BRCC36/C6.1A/CXorf53
Gene Symbols BRCC3
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 1-316 of human BRCC3.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79184
Test Specificity This antibody reacted with human BRCC3.
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.