Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BrdU, Mouse, Clone: 131-14871 (AH4H7-1), Abnova™

Catalog No. 89135537 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-537 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-537 Supplier Abnova Corporation Supplier No. MAB3900
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against BrdU.

Specifications

Antigen BrdU
Applications Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone 131-14871 (AH4H7-1)
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against BrdU.
Dilution The optimal working dilution should be determined by the end user.
Formulation In 10mM phosphate buffer, 150mM NaCl, pH 7.4 (0.09% sodium azide)
Host Species Mouse
Immunogen Bromodeoxyuridine (BrdU).
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Bromodeoxyuridine (BrdU), a thymidine analogue, is incorporated into DNA during replication. Anti-BrdU reacts with cells that incorporated BrdU during proliferation activity in a variety of specimens such as tumors, blood, cell lines and tissues.
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.