Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BRI3BP, Mouse, Clone: 3H1, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant BRI3BP.
Supplier: Abnova Corporation H00140707M01
Description
Sequence: MGARASGGPLARAGLLLLLLLLLLLGLLAPGAQGARGRGGAEKNSYRRTVNTFSQSVSSLFGEDNVRAAQKFLARLTERFVLGVDMFVETLWKVWTELLDVLGLDVSNLSQYFSPASVSSSPARALLLVGVVLLAYWFLSLTLGFTFSVLHVVFGRFFWIVRVVLFSMSCVYILHKYEGEPENAVLPLCFVVAVYFMTGPMGFYWRSSPSGPSNPSNPSVEEKLEHLEKQVRLLNIRLNRVLESLDRSKDKSpecifications
BRI3BP | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_080626.5 | |
BRI3BP | |
BRI3BP (NP_542193.3,1 a.a. ∽ 251 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human, Rat | |
Antibody | |
IgG2b κ |
ELISA, Western Blot | |
3H1 | |
Mouse monoclonal antibody raised against a full-length recombinant BRI3BP. | |
BRI3BP | |
BNAS1/HCCR-2/KG19 | |
Mouse | |
Protein A Purification | |
RUO | |
140707 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction