Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRUNOL6, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89107252 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-252 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-252 Supplier Abnova Corporation Supplier No. H00060677B01P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human BRUNOL6 protein.

Members of the CELF/BRUNOL protein family contain two N-terminal RNA recognition motif (RRM) domains, one C-terminal RRM domain, and a divergent segment of 160-230 aa between the second and third RRM domains. Members of this protein family regulate pre-mRNA alternative splicing and may also be involved in mRNA editing, and translation. [provided by RefSeq

Sequence: MTAAPGGSAQPAGPGPRLGFSTADSGVGMSGLNPGPAVPMKDHDAIKLFVGQIPRGLDEQDLKPLFEEFGRIYELTVLKDRLTGLHKGCAFLTYCARDSALKAQSALHEQKTLPGMNRPIQVKPAASEGRGEDRKLFVGMLGKQQGEEDVRRLFQPFGHIEECTVLRSPDGTSKGCAFVKFGSQGEAQAAIRGLHGSRTMAGASSSLVVKLADTDRERALRRMQQMAGHLGAFHPAPLPLGACGAYTTAILQHQAALLAAAQGPGLGPVAAVAAQMQHVAAFSLVAAPLLPAAAANSPPGSGPGTLPGLPAPIGVNGFGPLTPQTNGQPGSDTLYNNGLSPYPAQSPGVADPLQQAYAGMHHYAAAYPSAYAPVSTAFPQQPSALPQQQREGPEGCNLFIYHLPQEFGDAELIQTFLPFGAVVSAKVFVDRATNQSKCFGFVSFDNPTSAQTAIQAMNGFQIGMKRLKAQLKRPKDANRPY

Specifications

Antigen BRUNOL6
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human BRUNOL6 protein.
Formulation PBS with no preservative; pH 7.4
Gene BRUNOL6
Gene Accession No. BC030835
Gene Alias CELF6
Gene Symbols BRUNOL6
Host Species Mouse
Immunogen BRUNOL6 (AAH30835,1 a.a. ∽ 481 a.a) full-length human protein.
Purification Method Affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60677
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.