Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BSG, Mouse, Clone: MEM-M6/6, Abnova™

Catalog No. 89112872 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-872 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-872 Supplier Abnova Corporation Supplier No. MAB6448
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against BSG.

The protein encoded by this gene is a plasma membrane protein that is important in spermatogenesis, embryo implantation, neural network formation, and tumor progression. The encoded protein is also a member of the immunoglobulin superfamily. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Specifications

Antigen BSG
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone MEM-M6/6
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against BSG.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, 300mM NaCl, pH 7.4
Gene BSG
Gene Alias 5F7/CD147/EMMPRIN/M6/OK/TCSF
Gene Symbols BSG
Host Species Mouse
Immunogen CD147Rg, CH2-and CH3 domain of human IgG1 (CD147Rg is secreted by transfectants as a dimmer)., Protein A-CR purified soluble recombinant form of CD147, which consists of the cDNA coding for the hinge region
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 682
Test Specificity This antibody recognizes Ig domain D2 (membrane proximal) of CD147 (Neurothelin), a 50-60 KDa type I transmembrane glycoprotein primarily expressed on all leukocytes, red blood cells, platelets and endothelial cells; it is not expressed by resting lymphocytes.
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.