Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Mouse anti-BTLA/cd272, Clone: FLO67B,

Catalog No. MABF269MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF269MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF269MI Supplier MilliporeSigma™ Supplier No. MABF269
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects BTLA/cd272 clone: FLO67B in Human samples, and it is validated for Immunohistochemistry

The BTLA gene product B- and T-lymphocyte attenuator, also known as cluster of differentiation 272 (CD272), is a T cell coinhibitory receptor of the immunoglobulin superfamily with structural similarity to programmed cell death 1 (PD1) and CTLA4. BTLA is reported to negatively regulate immune responses via interaction with the B7 superfamily costimulatory molecule B7-H4 as well as with tumor necrosis factor receptor superfamily member 14 (TNFRSF14), also known as herpes virus entry mediator (HVEM). Polymorphisms in the BTLA gene have been associated with an increased risk of rheumatoid arthritis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BTLA/cd272
Applications Immunohistochemistry
Classification Monoclonal
Clone FLO67B
Concentration 1mg/mL
Formulation Purified mouse monoclonal IgG1λ in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. Q7Z6A9
Gene Symbols BTLA
Host Species Mouse
Immunogen Recombinant protein corresponding to human BTLA/cd272.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Inflammation and Immunology
Primary or Secondary Primary
Gene ID (Entrez) NP_001078826
Target Species Human
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Form Purified
Isotype IgG1 λ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.