Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bu-1, Mouse, Clone: 5-11G2, Abnova™
Mouse monoclonal antibody raised against Bu-1.
Supplier: Abnova Corporation MAB3279
Specifications
Bu-1 | |
Monoclonal | |
Unconjugated | |
Flow Cytometry (1 ug/106 cells) The optimal working dilution should be determined by the end user. | |
Mouse | |
500 μg | |
Primary | |
Chicken | |
Liquid |
Flow Cytometry, Immunofluorescence, Immunohistochemistry (Frozen) | |
5-11G2 | |
Mouse monoclonal antibody raised against Bu-1. | |
In 100mM BBS, pH 8.2 (0.09% sodium azide) | |
Bursal cells from 2- to 5-week-old CHA (Bu-1b) strain chickens. | |
RUO | |
Chicken Bu-1b alloantigen, Mr 70 KDa. | |
Store at 4°C. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction