Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bu-1a, Mouse, PE, Clone: 21-1A4, Abnova™

Catalog No. 89136070 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-070 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-070 Supplier Abnova Corporation Supplier No. MAB5844
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native Bu-1a.

Specifications

Antigen Bu-1a
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation
Classification Monoclonal
Clone 21-1A4
Conjugate PE
Description Mouse monoclonal antibody raised against native Bu-1a.
Dilution Flow Cytometry (0.2 ug/106 cells) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Host Species Mouse
Immunogen Native purified from Bursal cells of one-day-old H.B15 (Bu-1 a/b) strain chickens.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Chicken Bu-1a alloantigen, Mr 70 KDa.
Target Species Chicken
Content And Storage Store in the dark at 4°C. Do not freeze.
Avoid prolonged exposure to light.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.