Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

c-Kit, Mouse, PE, Clone: kit2c75, Abnova™

Catalog No. 89136075 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-075 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-075 Supplier Abnova Corporation Supplier No. MAB5849
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against c-Kit.

Specifications

Antigen c-Kit
Applications Flow Cytometry
Classification Monoclonal
Clone kit2c75
Conjugate PE
Description Mouse monoclonal antibody raised against c-Kit.
Dilution Flow Cytometry (0.2 ug/106 cells) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Host Species Mouse
Immunogen Sp/chkit4A4, Sp2/0 cells, transfected with chicken c-kit cDNA.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Chicken c-Kit.
Target Species Chicken
Content And Storage Store in the dark at 4°C. Do not freeze.
Avoid prolonged exposure to light.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.