Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

c-MAF Mouse, PE, Clone: T54-853, BD

Catalog No. BDB565795 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB565795 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB565795 Supplier BD Biosciences Supplier No. 565795
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The T54-853 monoclonal antibody recognizes c-Maf, a transcription factor that is also known as, MAF, c-Maf protooncogene, or V-maf musculoaponeurotic fibrosarcoma oncogene homolog. c-Maf belongs to the basic leucine zipper (bZIP) family of transcription factors. c-Maf acts as a homodimer or can dimerize with other bZIP transcription factors to either activate or repress the expression of various genes. It plays multiple roles in the differentiation and effector functions of leucocytes, including macrophages, Th2 cells, Th17 cells, and T follicular helper (Tfh) cells. c-Maf drives the expression of IL-10 by activated macrophages, IL-21 by activated Tfh and Th17 cells, and IL-4 by Th2 cells.

TRUSTED_SUSTAINABILITY

Specifications

Antigen c-MAF
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone T54-853
Concentration 0.2mg/mL
Conjugate PE
Description MAF; v-MAF; MAF; AYGRP; CCA4; CTRCT21
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human c-MAF Recombinant Protein
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.