Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

c-Myc tag, Mouse, Clone: 289-19510, Abnova™

Catalog No. 89135635 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-635 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-635 Supplier Abnova Corporation Supplier No. MAB4164
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against synthetic peptides of c-Myc tag.

Specifications

Antigen c-Myc tag
Applications Immunohistochemistry (Frozen), Western Blot
Classification Monoclonal
Clone 289-19510
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic peptides of c-Myc tag.
Dilution The optimal working dilution should be determined by the end user.
Formulation In 10mM phosphate buffer, 150mM NaCl, pH 7.4 (0.09% sodium azide)
Host Species Mouse
Immunogen A synthetic peptide corresponding to c-Myc tag.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Myc tag (C-myc).
Content And Storage Store at 4°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.