Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C-peptide, Mouse, Clone: CC27, Abnova™

Catalog No. 89135255 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-255 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-255 Supplier Abnova Corporation Supplier No. MAB2818
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against synthetic peptide of C-peptide.

Specifications

Antigen C-peptide
Applications ELISA
Classification Monoclonal
Clone CC27
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic peptide of C-peptide.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Host Species Mouse
Immunogen A synthetic peptide corresponding to rat C-peptide.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody detects to rat C-peptide isoforms (rat C-peptide I and/or rat C-peptide II). Specific to C-terminal part of rat C-peptide.
Target Species Rat
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.