Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C14orf21, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89107665 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-665 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-665 Supplier Abnova Corporation Supplier No. H00161424B01P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human C14orf21 protein.

Sequence: MGQGPRSPHKVGRRFPAGGKRGRGAKGSGRPLPGRKRQPWPPPDGRSEPAPDSHPHLSPEALGYFRRALSALKEAPETGEERDLMVHNIMKEVETQALALSTNRTGSEMLQELLGFSPLKPLCRVWAALRSNLRTVACHRCGVHVLQSALLQLPRLLGSAAEEEEEEEEEDGKDGPTETLEELVLGLAAEVCDDFLVYCGDTHGSFVVRTLLQVLGGTILESERARPRGSQSSEAQKTPAQECKPADFEVPETFLNRLQDLSSSFLKDIAVFITDKISSFCLQVALQVLHRKLPQFCAHLCNAVIGYLNTRGSSVDGSPLLLFLRDQTSSRLLEQVLLVLEPPRLQSLFEEHLQGQLQTLAAHPIANFPLQRLLDAVTTPELLSPVFEELSPVLEAVLAQGHPGVVIALVGACRRVGAYQAKVLQLLLEAFHCAEPSSRQVACVPLFATLMAYEVYYGLTEEEGAVPAEHQVAMAAARALGDVTVLGSLLLQHLLHFSTPGLVLRSLGALTGPQLLSLAQSPGRKLLLSLGSRTRS

Specifications

Antigen C14orf21
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human C14orf21 protein.
Formulation PBS with no preservative; pH 7.4
Gene C14orf21
Gene Accession No. BC025332.1
Gene Alias KIAA2021
Gene Symbols C14orf21
Host Species Mouse
Immunogen C14orf21 (AAH25332.1,1 a.a. ∽ 536 a.a) full-length human protein.
Purification Method Affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 161424
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.