Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C1QA, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89104178 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-104-178 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-104-178 Supplier Abnova Corporation Supplier No. H00000712B02P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human C1QA protein.

This gene encodes a major constituent of the human complement subcomponent C1q. C1q associates with C1r and C1s in order to yield the first component of the serum complement system. Deficiency of C1q has been associated with lupus erythematosus and glomerulonephritis. C1q is composed of 18 polypeptide chains: six A-chains, six B-chains, and six C-chains. Each chain contains a collagen-like region located near the N terminus and a C-terminal globular region. The A-, B-, and C-chains are arranged in the order A-C-B on chromosome 1. This gene encodes the A-chain polypeptide of human complement subcomponent C1q. [provided by RefSeq

Sequence: MEGPRGWLVLCVLAISLASMVTEDLCRAPDGKKGEAGRPGRRGRPGLKGEQGEPGAPGIRTGIQGLKGDQGEPGPSGNPGKVGYPGPSGPLGARGIPGIKGTKGSPGNIKDQPRPAFSAIRRNPPMGGNVVIFDTVITNQEEPYQNHSGRFVCTVPGYYYFTFQVLSQWEICLSIVSSSRGQVRRSLGFCDTTNKGLFQVVSGGMVLQLQQGDQVWVEKDPKKGHIYQGSEADSVFSGFLIFPSA

Specifications

Antigen C1QA
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human C1QA protein.
Formulation PBS with no preservative; pH 7.4
Gene C1QA
Gene Accession No. NM_015991.2
Gene Symbols C1QA
Host Species Mouse
Immunogen C1QA (NP_057075.1,1 a.a. ∽ 245 a.a) full-length human protein.
Purification Method Affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 712
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.