Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C4A, Mouse, Clone: TDM16, Abnova™

Catalog No. 89109638 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-109-638 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-109-638 Supplier Abnova Corporation Supplier No. MAB11491

Mouse monoclonal antibody raised against C4A.

This gene encodes the acidic form of complement factor 4, part of the classical activation pathway. The protein is expressed as a single chain precursor which is proteolytically cleaved into a trimer of alpha, beta, and gamma chains prior to secretion. The trimer provides a surface for interaction between the antigen-antibody complex and other complement components. The alpha chain may be cleaved to release C4 anaphylatoxin, a mediator of local inflammation. Deficiency of this protein is associated with systemic lupus erythematosus and type I diabetes mellitus. This gene localizes to the major histocompatibility complex (MHC) class III region on chromosome 6. Varying haplotypes of this gene cluster exist, such that individuals may have 1, 2, or 3 copies of this gene. [provided by RefSeq

Specifications

Antigen C4A
Applications Immunofluorescence
Classification Monoclonal
Clone TDM16
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against C4A.
Dilution Immunofluorescence (0.5-1.0 ug/mL) The optimal working dilution should be determined by the end user.
Formulation 10mM PBS, 0.05% BSA, and 0.05% Azide
Gene C4A
Gene Alias C4/C4A2/C4A3/C4A4/C4A6/C4S/CO4/CPAMD2/MGC164979/RG
Gene Symbols C4A
Host Species Mouse
Immunogen Human C4A.
Purification Method Protein A/G purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 720
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.