Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C5AR1, Mouse, Clone: S5/1, Abnova™

Catalog No. 89112226 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-226 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-226 Supplier Abnova Corporation Supplier No. MAB5330
Add to Cart
Add to Cart
Request a Quote

Mouse monoclonal antibody raised against synthetic peptide of C5AR1.

Specifications

Antigen C5AR1
Applications Flow Cytometry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone S5/1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic peptide of C5AR1.
Dilution Flow Cytometry (10 ul of the suggested dilution (1:50-1:100) to label 5 x 105 cells in 100 ul) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene C5AR1
Gene Alias C5A/C5AR/C5R1/CD88
Gene Symbols C5AR1
Host Species Mouse
Immunogen A synthetic peptide corresponding to N-terminus extracellular domain (met1-Asn31) of human C5AR1.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 728
Test Specificity This antibody recognizes the C5a receptor (C5aR) CD88, which is predominantly expressed on cells of the myeloid lineage. Clone S5/1 was raised against a synthetic peptide comprising the Nterminal extracellular domain of the C5aR (met1-Asn31) and has recently been shown to recognize the heptameric peptide (D15DKDTLD21). Clone S5/1 has been shown to inhibit the binding of C5a to its receptor. Serotec recommend the use of the monoclonal antibody for functional studies. Cross reacts with human.
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.