Learn More
C6orf206, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human C6orf206 protein.
Supplier: Abnova Corporation H00221421B01P
Description
Radial spokes are regularly spaced along cilia, sperm, and flagella axonemes and have a multisubunit 'stalk' and 'head' that form a signal transduction scaffold between the central microtubule pair and dynein (see MIM 603297) arms. RSPH9 is predicted to be a component of the radial spoke head based on homology with proteins in the biflagellate alga Chlamydomonas reinhardtii and other ciliates (Castleman et al., 2009 [PubMed 19200523]).[supplied by OMIM
Sequence: MDADSLLLSLELASGSGQGLSPDRRASLLTSLMLVKRDYRYDRVLFWGRILGLVADYYIAQGLSEDQLAPRKTLYSLNCTEWSLLPPATEEMVAQSSVVKGRFMGDPSYEYEHTELQKVNEGEKVFEEEIVVQIKEETRLVSVIDQIDKAVAIIPRGALFKTPFGPTHVNRTFEGLSLSEAKKLSSYFHFREPVELKNKTLLEKADLDPSLDFMDSLEHDIPKGSWSIQMERGNALVVLRSLLWPGLTFYHAPRTKNYGYVYVGTGEKNMDLPFMLSpecifications
C6orf206 | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human C6orf206 protein. | |
C6orf206 | |
FLJ30845/MRPS18AL1 | |
Mouse | |
Affinity chromatography | |
RUO | |
221421 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Purified |
Immunofluorescence, Western Blot | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_152732 | |
C6orf206 | |
C6orf206 (NP_689945,1 a.a. ∽ 276 a.a) full-length human protein. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only