Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C7orf16, Mouse, Clone: 10H11, Abnova™

Catalog No. 89107817 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-817 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-817 Supplier Abnova Corporation Supplier No. MAB0006

Mouse monoclonal antibody raised against synthetic phosphopeptide of C7orf16.

Specifications

Antigen C7orf16
Applications ELISA, Western Blot
Classification Monoclonal
Clone 10H11
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic phosphopeptide of C7orf16.
Dilution Western Blot (1-2 ug/mL) ELISA (1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 10mM HEPES/KOH, 150mM NaCl, pH 7.5 (50% glycerol)
Gene C7orf16
Gene Accession No. AF097730
Gene Alias GSBS
Gene Symbols C7orf16
Host Species Mouse
Immunogen Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding T68/119 of human C7orf16.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10842
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.