Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CA 19-9 Mouse anti-Human, Monoclonal Antibody, Abnova™

Catalog No. 89134954 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-134-954 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-134-954 Supplier Abnova Corporation Supplier No. MAB1398
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native CA 19-9.

Specifications

Antigen CA 19-9
Applications Immunohistochemistry
Classification Monoclonal
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native CA 19-9.
Dilution The optimal working dilution should be determined by the end user.
Formulation In 15mM KPO4 buffer, 0.85% NaCl, pH 7.4 (0.09% sodium azide)
Host Species Mouse
Immunogen Native purified human CA 19-9.
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody is specific to CA 19-9. No cross-reactivity with human AFP, CEA, PAP, PSA, CA 125, or CA 15-3.
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.