Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CA19-9 (Lewis LeA), Mouse, Clone: Sla 203, Abnova™

Catalog No. 89136143 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-143 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-143 Supplier Abnova Corporation Supplier No. MAB6188
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against CA19-9 (Lewis LeA).

Specifications

Antigen CA19-9 (Lewis LeA)
Applications Immunocytochemistry, Immunohistochemistry
Classification Monoclonal
Clone Sla 203
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against CA19-9 (Lewis LeA).
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:50-1:100) Immunohistochemistry (Frozen sections) (1:50-1:100) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (1% BSA, 0.05% sodium azide)
Host Species Mouse
Immunogen Native purified from ovarian cyst.
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.