Learn More
CABC1, Mouse, Clone: 5A4, Abnova™
Mouse monoclonal antibody raised against a partial recombinant CABC1.
Supplier: Abnova Corporation H00056997M03A
Description
This gene encodes a mitochondrial protein similar to yeast ABC1, which functions in an electron-transferring membrane protein complex in the respiratory chain. It is not related to the family of ABC transporter proteins. Expression of this gene is induced by the tumor suppressor p53 and in response to DNA damage, and inhibiting its expression partially suppresses p53-induced apoptosis. Alternatively spliced transcript variants have been found; however, their full-length nature has not been determined. [provided by RefSeq
Sequence: MAAILGDTIMVAKGLVKLTQAAVETHLQHLGIGGELIMAARALQSTAVEQIGMFLGKVQGQDKHEEYFAENFGGPEGEFHFSVPHAAGASTDFSSASAPDSpecifications
CABC1 | |
Monoclonal | |
Unconjugated | |
ascites with no preservative | |
BC005171 | |
CABC1 | |
CABC1 (AAH05171,1 a.a. ∽ 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
RUO | |
56997 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Ascites |
ELISA, Western Blot | |
5A4 | |
Mouse monoclonal antibody raised against a partial recombinant CABC1. | |
CABC1 | |
ADCK3/ARCA2/COQ8/MGC4849/SCAR9 | |
Mouse | |
200 μL | |
Primary | |
Human | |
Antibody | |
IgM κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.