Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CABIN1, Mouse, Clone: 2F5, Abnova™

Catalog No. 89106617 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-106-617 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-106-617 Supplier Abnova Corporation Supplier No. H00023523M02A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant CABIN1.

Calcineurin plays an important role in the T-cell receptor-mediated signal transduction pathway. The protein encoded by this gene binds specifically to the activated form of calcineurin and inhibits calcineurin-mediated signal transduction. The encoded protein is found in the nucleus and contains a leucine zipper domain as well as several PEST motifs, sequences which confer targeted degradation to those proteins which contain them. At least four alternatively spliced transcripts have been found for this gene, but the full-length nature of most of them has not been determined. [provided by RefSeq

Sequence: MIRIAALNASSTIEDDHEGSFKSHKTQTKEAQEAEAFALYHKALDLQKHDRFEESAKAYHELLEASLLREAVSSGDEKEGLKHPGLILKYSTYKNLAQLAAQREDLETAM

Specifications

Antigen CABIN1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2F5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant CABIN1.
Formulation ascites with no preservative
Gene CABIN1
Gene Accession No. NM_012295
Gene Alias CAIN/KIAA0330/PPP3IN
Gene Symbols CABIN1
Host Species Mouse
Immunogen CABIN1 (NP_036427,1 a.a. ∽ 110 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23523
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.