Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cacna1g, Mouse, ATTO390, Clone: S178A-9, Abnova™

Catalog No. 89109699 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-109-699 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-109-699 Supplier Abnova Corporation Supplier No. MAB11549
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant of Cacna1g.

Specifications

Antigen Cacna1g
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone S178A-9
Conjugate Atto 390
Description Mouse monoclonal antibody raised against recombinant of Cacna1g.
Dilution Immunohistochemistry Western blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (50% glycerol)
Gene Cacna1g
Gene Alias Cav3.1d/[a]1G/a1G/alpha-1G/mKIAA1123
Gene Symbols Cacna1g
Host Species Mouse
Immunogen Recombinant fusion protien corresponding to amino acids 2052-2172 at C-terminus of mouse Cacna1g.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12291
Test Specificity Detects ∼<200 kDa. Does not cross-react with Cacna1h.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.