Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CALCA, Mouse, Clone: 3B1 1C10, Abnova™

Catalog No. 89111527 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-527 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-527 Supplier Abnova Corporation Supplier No. MAB3673
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against synthetic peptide of CALCA.

Calcitonin is a peptide hormone synthesized by the parafollicular cells of the thyroid. It causes reduction in serum calcium, an effect opposite to that of parathyroid hormone (PTH; MIM 168450).[supplied by OMIM

Specifications

Antigen CALCA
Applications ELISA
Classification Monoclonal
Clone 3B1 1C10
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic peptide of CALCA.
Dilution The optimal working dilution should be determined by the end user.
Formulation In BBS
Gene CALCA
Gene Alias CALC1/CGRP/CGRP-I/CGRP1/CT/KC/MGC126648
Gene Symbols CALCA
Host Species Mouse
Immunogen A synthetic peptide corresponding to human CALCA.
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 796
Test Specificity This antibody recognizes natural human CALCA.
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.