Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Calca, Mouse, Monoclonal Antibody, Abnova™
Mouse monoclonal antibody raised against native Calca.
Supplier: Abnova Corporation MAB8173
Specifications
Calca | |
Monoclonal | |
Mouse monoclonal antibody raised against native Calca. | |
In 10mM HEPES, 150mM NaCl, pH 7.5 (50% glycerol, 0.1mg/mL BSA) | |
CAL6/CGRP/Cal1/Calc/RATCAL6/calcitonin | |
Mouse | |
Protein G purification | |
RUO | |
24241 | |
Human, Rat | |
Liquid |
Immunocytochemistry | |
Unconjugated | |
Immunocytochemistry (1:50-1:100) The optimal working dilution should be determined by the end user. | |
Calca | |
Calca | |
Native purified rat Calca. | |
100 μL | |
Primary | |
This antibody has been directly tested for reactivity in rat and human. It is expected that This antibody will react with most mammals. specific to Calcitonin Gene Related Peptide. Also recognizes rat beta-CGRP and human alpha-CGRP (10-fold less sensitivity compared to rat alpha-CGRP). Suggested positive controls are dorsal root ganglia, pancreas and gut. | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction