Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Caldesmon, Mouse, Clone: 12B5, Abnova™

Catalog No. 89135223 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-223 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-223 Supplier Abnova Corporation Supplier No. MAB2773

Mouse monoclonal antibody raised against Caldesmon.

Specifications

Antigen Caldesmon
Applications ELISA, Western Blot
Classification Monoclonal
Clone 12B5
Conjugate Unconjugated
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Host Species Mouse
Immunogen Duck gizzard caldesmon (heavy chain).
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody reacts with heavy and light forms of caldesmon. No cross-react with other proteins.
Target Species Chicken, Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.