Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Calnexin Antibody (1C2.2D11) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$207.00 - $426.00
Specifications
| Antigen | Calnexin |
|---|---|
| Clone | 1C2.2D11 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot, Immunohistochemistry 7 ug/ml, Immunocytochemistry/Immunofluorescence 10 ug/ml, Immunohistochemistry-Paraffin 7 ug/ml, Flow (Intracellular) 5 ug/million cells |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP236570SS
![]() |
Novus Biologicals
NBP236570SS |
0.025 mg |
Each for $207.00
|
|
|||||
NBP236570
![]() |
Novus Biologicals
NBP236570 |
0.1 mg |
Each for $426.00
|
|
|||||
Description
Calnexin Monoclonal specifically detects Calnexin in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Intracellular).Specifications
| Calnexin | |
| 1.0 mg/mL | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| ER Markers, Neuroscience, Phospho Specific, Sensory Systems, Unfolded Protein Response, Vision | |
| PBS with 0.05% Sodium Azide | |
| calnexin, CNX, IP90FLJ26570, Major histocompatibility complex class I antigen-binding protein p88, P90 | |
| CANX | |
| IgG2b κ | |
| Protein G purified | |
| Immunogen's sequence similarity with other species: Mouse (88%), Duck (81%), Xenopus (76%) |
| 1C2.2D11 | |
| Western Blot, Immunohistochemistry 7 ug/ml, Immunocytochemistry/Immunofluorescence 10 ug/ml, Immunohistochemistry-Paraffin 7 ug/ml, Flow (Intracellular) 5 ug/million cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| P27824 | |
| 821 | |
| Partial recombinant human Calnexin protein (between amino acids 1-300). [UniProt P27824] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title