Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CAMK2G, Mouse, Clone: 22B1, Abnova™

Catalog No. 89136451 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-451 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-451 Supplier Abnova Corporation Supplier No. MAB8281
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against synthetic phosphopeptide of CAMK2G.

Specifications

Antigen CAMK2G
Applications Immunofluorescence, Western Blot
Classification Monoclonal
Clone 22B1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic phosphopeptide of CAMK2G.
Dilution Western Blot (1 ug/mL) Immunofluorescence (10 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (50% glycerol, 0.09% sodium azide)
Host Species Mouse
Immunogen Synthetic phosphopeptide corresponding to amino acids 281-294 residues surrounding T286 of CAMK2G.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody recognizes the phosphorylated form of CAMK2G. It does not react with nonphosphorylated form.
Target Species Mouse, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.