Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CaMKII, Mouse, Clone: 22B1, Abnova™
Mouse monoclonal antibody raised against synthetic peptide of CaMKII.
Supplier: Abnova Corporation MAB6627
Specifications
CaMKII | |
Monoclonal | |
Unconjugated | |
Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user. | |
Mouse | |
100 μg | |
Primary | |
Rat | |
Liquid |
ELISA, Immunofluorescence, Immunoprecipitation, Western Blot | |
22B1 | |
Mouse monoclonal antibody raised against synthetic peptide of CaMKII. | |
In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide) | |
A synthetic peptide corresponding to CaMKII. | |
RUO | |
Detects phosphorylated CaMKII from rat tissues. This antibody is specific to alpha and beta subunits of CaMKII only when they are phosphorylated at Thr-286/287 (in beta). | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction